Learn More
Biomatik Corporation Phospho-HDAC1 (Ser421) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51236
Encompass_Preferred
Phospho-HDAC1 (Ser421) Colorimetric Cell-Based ELISA Kit for detecting Phospho-HDAC1 (Ser421) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13547. Gene ID: 3065. Target Synonyms: HD1; Histone deacetylase 1; RPD3L1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1410