Learn More
Biomatik Corporation Phospho-PKC α (Tyr657) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51239
Encompass_Preferred
Phospho-PKC α (Tyr657) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PKC α (Tyr657) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P17252. Gene ID: 5578. Target Synonyms: kinase PKC-alpha; KPCA; PKC III; PKC-A; PKC-alpha; PKC-III; PRKACA; PRKCA; Protein kinase C; alpha type. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1412