Learn More
Biomatik Corporation Phospho-PKC θ (Ser695) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51242
Encompass_Preferred
Phospho-PKC θ (Ser695) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PKC θ (Ser695) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q04759. Gene ID: 5588. Target Synonyms: KPCT; PKC-theta; PKCQ; PRKCQ; PRKCT; Protein kinase C; theta type; kinase PKC-theta; nPKC-theta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1414