Learn More
Biomatik Corporation Phospho-APC (Ser2054) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51252
Encompass_Preferred
Phospho-APC (Ser2054) Colorimetric Cell-Based ELISA Kit for detecting Phospho-APC (Ser2054) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P25054. Gene ID: 324. Target Synonyms: Adenomatous polyposis coli protein; adenomatous polyposis of the colon gene; APC; APC protein; DP2.5; mAPC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1424