Learn More
Biomatik Corporation Phospho-ERK8 (Thr175+Tyr177) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51298
Encompass_Preferred
Phospho-ERK8 (Thr175+Tyr177) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ERK8 (Thr175+Tyr177) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8TD08. Gene ID: 225689. Target Synonyms: Extracellular signal-regulated kinase 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1465