Learn More
Biomatik Corporation Phospho-HDAC3 (Ser424) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51303
Encompass_Preferred
Phospho-HDAC3 (Ser424) Colorimetric Cell-Based ELISA Kit for detecting Phospho-HDAC3 (Ser424) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O15379. Gene ID: 8841. Target Synonyms: HD3; Histone deacetylase 3; RPD3-2; SMAP45. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1470