Learn More
Biomatik Corporation Phospho-Caspase 1 (Ser376) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51326
Encompass_Preferred
Phospho-Caspase 1 (Ser376) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Caspase 1 (Ser376) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P29466. Gene ID: 834. Target Synonyms: CASP-1; caspase-1; ICE; IL-1 beta converting enzyme; IL-1BC; IL1BCE; interleukin-1 beta convertase; interleukin-1 beta converting enzyme; p45. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1490