Learn More
Biomatik Corporation Phospho-hnRNP C1/2 (Ser260) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291496
Encompass_Preferred
Phospho-hnRNP C1/2 (Ser260) Colorimetric Cell-Based ELISA Kit for detecting Phospho-hnRNP C1/2 (Ser260) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07910. Gene ID: 3183. Target Synonyms: Heterogeneous nuclear ribonucleoproteins C1/C2; hnRNP C1/hnRNP C2; HNRNPC; HNRPC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1496 Supplier Biomatik Corporation Supplier No. EKA51332
Phospho-hnRNP C1/2 (Ser260) Colorimetric Cell-Based ELISA Kit for detecting Phospho-hnRNP C1/2 (Ser260) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07910. Gene ID: 3183. Target Synonyms: Heterogeneous nuclear ribonucleoproteins C1/C2; hnRNP C1/hnRNP C2; HNRNPC; HNRPC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.