Learn More
Biomatik Corporation Phospho-GR (Ser211) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51021
Encompass_Preferred
Phospho-GR (Ser211) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GR (Ser211) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04150. Gene ID: 2908. Target Synonyms: GCR; GRL; GRL1; NR3C1; glucocorticoid receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1227