Learn More
Biomatik Corporation Phospho-Catenin-β (Ser37) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51403
Encompass_Preferred
Phospho-Catenin-β (Ser37) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Catenin-β (Ser37) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35222. Gene ID: 1499. Target Synonyms: Catenin beta-1; Beta-catenin; CTNNB1; CTNNB; OK/SW-cl.35; PRO2286. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9051