Learn More
Biomatik Corporation Phospho-FOXO1A (Ser256) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51453
Encompass_Preferred
Phospho-FOXO1A (Ser256) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FOXO1A (Ser256) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q12778. Gene ID: 2308. Target Synonyms: FKHR; forkhead box O1A (FKHR) Forkhead box protein O1A; Forkhead in rhabdomyosarcoma; FOXO1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9064