Learn More
Biomatik Corporation Phospho-STAT2 (Tyr690) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51037
Encompass_Preferred
Phospho-STAT2 (Tyr690) Colorimetric Cell-Based ELISA Kit for detecting Phospho-STAT2 (Tyr690) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52630. Gene ID: 6773. Target Synonyms: signal transducer and activator of transcription 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1241