Learn More
Biomatik Corporation Phospho-WNK1 (Thr58) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291244
Encompass_Preferred
Phospho-WNK1 (Thr58) Colorimetric Cell-Based ELISA Kit for detecting Phospho-WNK1 (Thr58) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9H4A3. Gene ID: 65125. Target Synonyms: WNK lysine deficient protein kinase 1; KDP; PRKWNK1; PROTEIN KINASE; LYSINE-DEFICIENT 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1244 Supplier Biomatik Corporation Supplier No. EKA51041
Phospho-WNK1 (Thr58) Colorimetric Cell-Based ELISA Kit for detecting Phospho-WNK1 (Thr58) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9H4A3. Gene ID: 65125. Target Synonyms: WNK lysine deficient protein kinase 1; KDP; PRKWNK1; PROTEIN KINASE; LYSINE-DEFICIENT 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.