Learn More
Biomatik Corporation Phospho-ATF1 (Ser63) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51053
Encompass_Preferred
Phospho-ATF1 (Ser63) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ATF1 (Ser63) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P18846. Gene ID: 466. Target Synonyms: ATF1; Activating transcription factor 1; Cyclic-AMP-dependent transcription factor ATF-1; TCR-ATF1; TREB36 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1256