Learn More
Biomatik Corporation Phospho-Csk (Ser364) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291258
Encompass_Preferred
Phospho-Csk (Ser364) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Csk (Ser364) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P41240. Gene ID: 1445. Target Synonyms: C-SRC kinase; Protein-tyrosine kinase CYL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1258 Supplier Biomatik Corporation Supplier No. EKA51057
Phospho-Csk (Ser364) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Csk (Ser364) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P41240. Gene ID: 1445. Target Synonyms: C-SRC kinase; Protein-tyrosine kinase CYL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.