Learn More
Biomatik Corporation Phospho-Dynamin-1 (Ser774) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51062
Encompass_Preferred
Phospho-Dynamin-1 (Ser774) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Dynamin-1 (Ser774) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q05193. Gene ID: 1759. Target Synonyms: B-dynamin; D100; DNM; DNM1; Dynamin BREDNM19; Dynamin brain; dynamin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1263