Learn More
Biomatik Corporation Phospho-IL-3Rβ (Tyr593) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51080
Encompass_Preferred
Phospho-IL-3Rβ (Tyr593) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IL-3Rβ (Tyr593) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P32927. Gene ID: 1439. Target Synonyms: AIC2B; CD131; CDw131; CSF2RB; CSF2RB1; CYRB; Cytokine receptor common beta chain precursor; IL-5R-B; IL3RB; IL3RB1; IL5RB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1279