Learn More
Biomatik Corporation Phospho-Ku80/XRCC5 (Thr714) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51085
Encompass_Preferred
Phospho-Ku80/XRCC5 (Thr714) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Ku80/XRCC5 (Thr714) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P13010. Gene ID: 7520. Target Synonyms: 86 kDa subunit of Ku antigen; ATP-dependent DNA helicase II; 80 kDa subunit; CTC box binding factor 85 kDa subunit; CTC85; CTCBF; DNA-repair protein XRCC5; G22P2; KU86; Lupus Ku autoantigen protein p86; Nuclear factor IV; Thyroid- lupus autoantigen; TLAA; XRCC5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1284