Learn More
Biomatik Corporation Phospho-KIT (Tyr703) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51117
Encompass_Preferred
Phospho-KIT (Tyr703) Colorimetric Cell-Based ELISA Kit for detecting Phospho-KIT (Tyr703) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10721. Gene ID: 3815. Target Synonyms: SCFR; Proto-oncogene tyrosine-protein kinase Kit; c-kit; CD117 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1312