Learn More
Biomatik Corporation Phospho-NFAT4 (Ser165) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291320
Encompass_Preferred
Phospho-NFAT4 (Ser165) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NFAT4 (Ser165) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q12968. Gene ID: 4775. Target Synonyms: Nuclear factor of activated T-cells cytoplasmic 3; NF-ATc3; NFATc3; T-cell transcription factor NFAT4; NF-AT4; NFATx; NFATC3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1320 Supplier Biomatik Corporation Supplier No. EKA51128
Phospho-NFAT4 (Ser165) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NFAT4 (Ser165) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q12968. Gene ID: 4775. Target Synonyms: Nuclear factor of activated T-cells cytoplasmic 3; NF-ATc3; NFATc3; T-cell transcription factor NFAT4; NF-AT4; NFATx; NFATC3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.