Learn More
Biomatik Corporation Phospho-Tuberin/TSC2 (Ser939) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291353
Encompass_Preferred
Phospho-Tuberin/TSC2 (Ser939) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Tuberin/TSC2 (Ser939) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49815. Gene ID: 7249. Target Synonyms: TSC2; tuberous sclerosis 2 gene; Tuberous sclerosis 2 homolog protein; Tuberous sclerosis 2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1353 Supplier Biomatik Corporation Supplier No. EKA51168
Phospho-Tuberin/TSC2 (Ser939) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Tuberin/TSC2 (Ser939) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49815. Gene ID: 7249. Target Synonyms: TSC2; tuberous sclerosis 2 gene; Tuberous sclerosis 2 homolog protein; Tuberous sclerosis 2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.