Learn More
Biomatik Corporation Phospho-ATP-Citrate Lyase (Ser454) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51181
Encompass_Preferred
Phospho-ATP-Citrate Lyase (Ser454) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ATP-Citrate Lyase (Ser454) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P53396. Gene ID: 47. Target Synonyms: ATP-citrate (pro-S-)-lyase; ATP-citrate lyase; Citrate cleavage enzyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1363