Learn More
Biomatik Corporation Phospho-CD32 (Tyr292) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51184
Encompass_Preferred
Phospho-CD32 (Tyr292) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CD32 (Tyr292) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P31994. Gene ID: 2213. Target Synonyms: CD32; CDW32; FC gamma receptor IIB; FC-gamma RII; FC-gamma RII-B; FC-gamma-RIIB; FCG2; FCG2B; FCGR2; FCGR2B; FCRII; FCgammaRIIB; IGFR2; IGG FC receptor II beta; IGG FC receptor II-B; LY-17; Low affinity immunoglobulin gamma FC region receptor II precursor; Low affinity immunoglobulin gamma FC region receptor II-B precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1365