Learn More
Biomatik Corporation Phospho-Chk1 (Ser296) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51186
Encompass_Preferred
Phospho-Chk1 (Ser296) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Chk1 (Ser296) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O14757. Gene ID: 1111. Target Synonyms: CHEK1; Chk1; kinase Chk1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1367