Learn More
Biomatik Corporation Phospho-PKR (Thr446) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51548
Encompass_Preferred
Phospho-PKR (Thr446) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PKR (Thr446) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P19525. Gene ID: 5610. Target Synonyms: Interferon-inducible RNA-dependent protein kinase; Eukaryotic translation initiation factor 2-alpha kinase 2; eIF-2A protein kinase 2; Protein kinase RNA-activated; PKR; p68 kinase; P1/eIF-2A protein kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9075