Learn More
Biomatik Corporation Phospho-PAK2 (Ser20) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291385
Encompass_Preferred
Phospho-PAK2 (Ser20) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PAK2 (Ser20) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13177. Gene ID: 5062. Target Synonyms: p21-activated kinase 2; PAK-2; Gamma-PAK; PAK65; S6/H4 kinase; p58. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1385 Supplier Biomatik Corporation Supplier No. EKA51207
Phospho-PAK2 (Ser20) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PAK2 (Ser20) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13177. Gene ID: 5062. Target Synonyms: p21-activated kinase 2; PAK-2; Gamma-PAK; PAK65; S6/H4 kinase; p58. Restrictions: For Research Use Only. Not for use in diagnostic procedures.