Learn More
Biomatik Corporation Phospho-ZAP-70 (Tyr493) Colorimetric Cell-Based ELISA Kit
Catalog No. 502289085
Encompass_Preferred
Phospho-ZAP-70 (Tyr493) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ZAP-70 (Tyr493) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P43403. Gene ID: 7535. Target Synonyms: 70 kDa zeta-associated protein; SRK; Syk-related tyrosine kinase; ZA70; ZAP-70; ZAP70; kinase ZAP70. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9085 Supplier Biomatik Corporation Supplier No. EKA51593
Phospho-ZAP-70 (Tyr493) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ZAP-70 (Tyr493) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P43403. Gene ID: 7535. Target Synonyms: 70 kDa zeta-associated protein; SRK; Syk-related tyrosine kinase; ZA70; ZAP-70; ZAP70; kinase ZAP70. Restrictions: For Research Use Only. Not for use in diagnostic procedures.