Learn More
Biomatik Corporation Phospho-CDC25A (Ser124) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291443
Encompass_Preferred
Phospho-CDC25A (Ser124) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDC25A (Ser124) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P30304. Gene ID: 993. Target Synonyms: CDC25M3; Dual specificity phosphatase Cdc25A; M-phase inducer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1443 Supplier Biomatik Corporation Supplier No. EKA51272
Phospho-CDC25A (Ser124) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDC25A (Ser124) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P30304. Gene ID: 993. Target Synonyms: CDC25M3; Dual specificity phosphatase Cdc25A; M-phase inducer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.