Learn More
Biomatik Corporation Phospho-Cyclin H (Thr315) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291449
Encompass_Preferred
Phospho-Cyclin H (Thr315) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Cyclin H (Thr315) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P51946. Gene ID: 902. Target Synonyms: CCNH; MO15-associated protein; p34; p37. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1449 Supplier Biomatik Corporation Supplier No. EKA51281
Phospho-Cyclin H (Thr315) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Cyclin H (Thr315) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P51946. Gene ID: 902. Target Synonyms: CCNH; MO15-associated protein; p34; p37. Restrictions: For Research Use Only. Not for use in diagnostic procedures.