Learn More
Biomatik Corporation Phospho-CaMK2-β/γ/δ (Thr287) Colorimetric Cell-Based ELISA Kit
Catalog No. 502289089
Encompass_Preferred
Phospho-CaMK2-β/γ/δ (Thr287) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CaMK2-β/γ/δ (Thr287) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13554; Q13555; Q13557. Gene ID: 816; 818; 817. Target Synonyms: CaM-kinase II beta chain; CaM kinase II subunit beta; CaMK-II subunit beta/ CaM-kinase II gamma chain; CaM kinase II subunit gamma; CaMK-II subunit gamma/ CaM-kinase II delta chain; CaM kinase II subunit delta; CaMK-II subunit delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9089 Supplier Biomatik Corporation Supplier No. EKA51620
Phospho-CaMK2-β/γ/δ (Thr287) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CaMK2-β/γ/δ (Thr287) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13554; Q13555; Q13557. Gene ID: 816; 818; 817. Target Synonyms: CaM-kinase II beta chain; CaM kinase II subunit beta; CaMK-II subunit beta/ CaM-kinase II gamma chain; CaM kinase II subunit gamma; CaMK-II subunit gamma/ CaM-kinase II delta chain; CaM kinase II subunit delta; CaMK-II subunit delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.