Learn More
Biomatik Corporation Phospho-DAPP1 (Tyr139) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291457
Encompass_Preferred
Phospho-DAPP1 (Tyr139) Colorimetric Cell-Based ELISA Kit for detecting Phospho-DAPP1 (Tyr139) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UN19. Gene ID: 27071. Target Synonyms: 3' phosphoinositide-interacting SH2 domain containing protein; B lymphocyte adapter protein BAM32; Bam32; Dual adaptor for phosphotyrosine and 3-phosphoinositides 1; PHISH; dual adapter for phosphotyrosine and 3-phosphoinositides. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1457 Supplier Biomatik Corporation Supplier No. EKA51289
Phospho-DAPP1 (Tyr139) Colorimetric Cell-Based ELISA Kit for detecting Phospho-DAPP1 (Tyr139) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UN19. Gene ID: 27071. Target Synonyms: 3' phosphoinositide-interacting SH2 domain containing protein; B lymphocyte adapter protein BAM32; Bam32; Dual adaptor for phosphotyrosine and 3-phosphoinositides 1; PHISH; dual adapter for phosphotyrosine and 3-phosphoinositides. Restrictions: For Research Use Only. Not for use in diagnostic procedures.