Learn More
Biomatik Corporation Phospho-MCM4 (Ser54) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51291
Encompass_Preferred
Phospho-MCM4 (Ser54) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MCM4 (Ser54) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P33991. Gene ID: 4173. Target Synonyms: CDC21 homolog; DNA replication licensing factor MCM4; P1-CDC21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1459