Learn More
Biomatik Corporation Phospho-GRK1 (Ser21) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51302
Encompass_Preferred
Phospho-GRK1 (Ser21) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GRK1 (Ser21) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15835. Gene ID: 6011. Target Synonyms: RHOK; RK; Rhodopsin kinase; kinase GRK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1469