Learn More
Biomatik Corporation Phospho-RFA2 (Thr21) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51313
Encompass_Preferred
Phospho-RFA2 (Thr21) Colorimetric Cell-Based ELISA Kit for detecting Phospho-RFA2 (Thr21) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15927. Gene ID: 6118. Target Synonyms: REPA2; RF-A; RP-A; RPA2; RPA32; replication factor-A protein 2; replication protein A 32 kDa subunit; replication protein A2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1478