Learn More
Biomatik Corporation Phospho-hnRPD (Ser83) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291498
Encompass_Preferred
Phospho-hnRPD (Ser83) Colorimetric Cell-Based ELISA Kit for detecting Phospho-hnRPD (Ser83) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14103. Gene ID: 3184. Target Synonyms: ARE element RNA-binding protein AUF1; AU-rich element RNA-binding protein 1; AUF1; HNRPD; Heterogeneous nuclear ribonucleoprotein D; Heterogeneous nuclear ribonucleoprotein D0; ROD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1498 Supplier Biomatik Corporation Supplier No. EKA51334
Phospho-hnRPD (Ser83) Colorimetric Cell-Based ELISA Kit for detecting Phospho-hnRPD (Ser83) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14103. Gene ID: 3184. Target Synonyms: ARE element RNA-binding protein AUF1; AU-rich element RNA-binding protein 1; AUF1; HNRPD; Heterogeneous nuclear ribonucleoprotein D; Heterogeneous nuclear ribonucleoprotein D0; ROD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.