Learn More
Biomatik Corporation Phospho-IL-4R/CD124 (Tyr497) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51341
Encompass_Preferred
Phospho-IL-4R/CD124 (Tyr497) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IL-4R/CD124 (Tyr497) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P24394. Gene ID: 3566. Target Synonyms: 582J2.1; CD124 antigen; IL-4R-alpha; IL4RA; Interleukin-4 receptor alpha chain precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1505