Learn More
Biomatik Corporation Phospho-Moesin/Ezrin/Radixin (Thr558) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51349
Encompass_Preferred
Phospho-Moesin/Ezrin/Radixin (Thr558) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Moesin/Ezrin/Radixin (Thr558) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P26038; P35241; P15311. Gene ID: 4478; 5962. Target Synonyms: MOES; MSN; Membrane-organizing extension spike protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1512