Learn More
Biomatik Corporation Phospho-CAMK2A/CAMK2D (Thr286) Colorimetric Cell-Based ELISA Kit
Catalog No. 502289052
Encompass_Preferred
Phospho-CAMK2A/CAMK2D (Thr286) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CAMK2A/CAMK2D (Thr286) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UQM7; Q13557. Gene ID: 815; 817. Target Synonyms: CAMK2A; CaM-kinase II alpha chain; CaMK-II alpha subunit; CaMKII-alpha; Calcium/calmodulin-dependent protein kinase type II alpha chain; KCC2A; kinase CaMK2-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-228-9052 Supplier Biomatik Corporation Supplier No. EKA51409
Phospho-CAMK2A/CAMK2D (Thr286) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CAMK2A/CAMK2D (Thr286) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UQM7; Q13557. Gene ID: 815; 817. Target Synonyms: CAMK2A; CaM-kinase II alpha chain; CaMK-II alpha subunit; CaMKII-alpha; Calcium/calmodulin-dependent protein kinase type II alpha chain; KCC2A; kinase CaMK2-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.