Learn More
Biomatik Corporation Phospho-4E-BP1 (Thr36) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291546
Encompass_Preferred
Phospho-4E-BP1 (Thr36) Colorimetric Cell-Based ELISA Kit for detecting Phospho-4E-BP1 (Thr36) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13541. Gene ID: 1978. Target Synonyms: 4EBP1; EIF4EBP1; Eukaryotic translation initiation factor 4E binding protein 1; Insulin-stimulated EIF-4E binding protein PHAS-I; P/OKCL.6; PHAS-1; PHAS-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1546 Supplier Biomatik Corporation Supplier No. EKA51388
Phospho-4E-BP1 (Thr36) Colorimetric Cell-Based ELISA Kit for detecting Phospho-4E-BP1 (Thr36) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13541. Gene ID: 1978. Target Synonyms: 4EBP1; EIF4EBP1; Eukaryotic translation initiation factor 4E binding protein 1; Insulin-stimulated EIF-4E binding protein PHAS-I; P/OKCL.6; PHAS-1; PHAS-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.