Learn More
Biomatik Corporation Phospho-AMPK1 (Ser485) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51392
Encompass_Preferred
Phospho-AMPK1 (Ser485) Colorimetric Cell-Based ELISA Kit for detecting Phospho-AMPK1 (Ser485) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13131. Gene ID: 5562. Target Synonyms: 5'-AMP-activated protein kinase catalytic alpha-1 chain; AAPK1; acetyl-CoA carboxylase kinase; AMPK alpha-1 chain; AMPK-alpha1; HMG-CoA redustase kinase; hormone-sensitive lipase kinase; kinase AMPK-alpha1; PRKAA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1549