Learn More
Biomatik Corporation Phospho-Estrogen Receptor-α (Ser106) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51445
Encompass_Preferred
Phospho-Estrogen Receptor-α (Ser106) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Estrogen Receptor-α (Ser106) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P03372. Gene ID: 2099. Target Synonyms: ER; ESR; ESR1; ESTR; ESTRA; Estradiol receptor; Estrogen receptor; NR3A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1589