Learn More
Biomatik Corporation Phospho-GluR1 (Ser863) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51461
Encompass_Preferred
Phospho-GluR1 (Ser863) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GluR1 (Ser863) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42261. Gene ID: 2890. Target Synonyms: Glutamate receptor 1; GluR-1; GluR-A; GluR-K1; Glutamate receptor ionotropic AMPA 1; AMPA-selective glutamate receptor 1; GRIA1; GLUH1; GLUR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1602