Learn More
Biomatik Corporation Phospho-GSK3α (Ser21) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51463
Encompass_Preferred
Phospho-GSK3α (Ser21) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GSK3α (Ser21) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49840. Gene ID: 2931. Target Synonyms: Factor A; GSK-3 alpha; GSK3-alpha; Glycogen synthase kinase-3 alpha; kinase GSK3-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1604