Learn More
Biomatik Corporation Phospho-IκB-α (Ser32/Ser36) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51485
Encompass_Preferred
Phospho-IκB-α (Ser32/Ser36) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IκB-α (Ser32/Ser36) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P25963. Gene ID: 4792. Target Synonyms: I-kappa-B-alpha; IkappaBalpha; IKBA; MAD3; Major histocompatibility complex enhancer-binding protein MAD3; NF-kappaB inhibitor alpha; NFKBI; NFKBIA; RL/IF-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1622