Learn More
Biomatik Corporation Phospho-Synuclein-α (Tyr136) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51578
Encompass_Preferred
Phospho-Synuclein-α (Tyr136) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Synuclein-α (Tyr136) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P37840. Gene ID: 6622. Target Synonyms: Alpha-synuclein; Non-A beta component of AD amyloid; Non-A4 component of amyloid precursor; NACP; SNCA; NACP; PARK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1704