Learn More
Biomatik Corporation Phospho-Casein Kinase I α (Tyr294) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51621
Encompass_Preferred
Phospho-Casein Kinase I α (Tyr294) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Casein Kinase I α (Tyr294) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P48729; Q8N752. Gene ID: 1452; 122011. Target Synonyms: CK1-alpha; CKI-alpha; CSNK1A1; Casein kinase I; alpha isoform; KC1A; kinase CK1-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1737