Learn More
Biomatik Corporation Phospho-PAK1 (Ser204) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51635
Encompass_Preferred
Phospho-PAK1 (Ser204) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PAK1 (Ser204) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13153. Gene ID: 5058. Target Synonyms: Serine/threonine-protein kinase PAK 1; p21-activated kinase 1; PAK-1; p65-PAK; Alpha-PAK; PAK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1749