Learn More
Biomatik Corporation Phospho-RSK1/2/3/4 (Ser221/227/S218/232) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51644
Encompass_Preferred
Phospho-RSK1/2/3/4 (Ser221/227/S218/232) Colorimetric Cell-Based ELISA Kit for detecting Phospho-RSK1/2/3/4 (Ser221/227/S218/232) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15418; P51812; Q15349; Q9UK32. Gene ID: 6195; 6197; 6196; 27330. Target Synonyms: S6K-alpha 1; 90 kDa ribosomal protein S6 kinase 1; p90-RSK 1; Ribosomal S6 kinase 1; RSK-1; pp90RSK1; p90S6K; MAP kinase-activated protein kinase 1a; MAPKAPK1A/ S6K-alpha 3; 90 kDa ribosomal protein S6 kinase 3; p90-RSK 3; Ribosomal S6 kinase 2; RSK-2; pp90RSK2; Insulin-stimulated protein kinase 1; ISPK-1; MAP kinase-activated protein kinase 1b; MAPKAPK1B/ S6K-alpha 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1754