Learn More
Biomatik Corporation Phospho-STAT5A (Ser780) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291696
Encompass_Preferred
Phospho-STAT5A (Ser780) Colorimetric Cell-Based ELISA Kit for detecting Phospho-STAT5A (Ser780) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42229. Gene ID: 6776. Target Synonyms: MGF; MPF; Mammary gland factor; STA5A; STAT5; STAT5A; Signal transducer and activator of transcription 5A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1696 Supplier Biomatik Corporation Supplier No. EKA51569
Phospho-STAT5A (Ser780) Colorimetric Cell-Based ELISA Kit for detecting Phospho-STAT5A (Ser780) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42229. Gene ID: 6776. Target Synonyms: MGF; MPF; Mammary gland factor; STA5A; STAT5; STAT5A; Signal transducer and activator of transcription 5A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.