Learn More
Biomatik Corporation Phospho-Caspase 8 (Tyr380) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51596
Encompass_Preferred
Phospho-Caspase 8 (Tyr380) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Caspase 8 (Tyr380) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14790. Gene ID: 841. Target Synonyms: Apoptotic cysteine protease; Apoptotic protease Mch-5; CAP4; Caspase-8 precursor; FADD-homologous ICE/CED-3- like protease; FADD-like ICE; FLICE; ICE-like apoptotic protease 5; ICE8; MACH; MCH5; MORT1-associated CED-3 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1716